Assay Details
| Gene Symbol: Pam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025571.1 | NP_079847.1 | 5 | 499 | 144 | |
| GenBank mRNA | AK008380.1 | - | 5 | 499 | 144 | |
| BC096419.1 | - | 5 | 488 | 1546951 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511148 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511152 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166543 | AK003227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166543 | AK008380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166543 | BC024346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166543 | BC096419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166543 | NM_025571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120558 | AK008380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120558 | BC096419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120558 | NM_025571.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229268 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143839 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120558 | AK008380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120558 | BC096419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120558 | NM_025571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166543 | AK003227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166543 | AK008380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166543 | BC024346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166543 | BC096419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166543 | NM_025571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246944 | AF349454.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246944 | AK003227.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246944 | AK008380.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246944 | BC024346.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246944 | BC096419.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511160 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143839 | - | - |
| Whole Mouse Genome G4122A | A_51_P246944 | AF349454.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246944 | AK003227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246944 | AK008380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246944 | BC024346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246944 | BC096419.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120558 | AK008380.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120558 | BC096419.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120558 | NM_025571.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136897 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143839 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12908680 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12523400 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12908674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12908671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12908676 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511148 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12784368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12753431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12908677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13031135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12908674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12753433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12625269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12878842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12587082 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10362894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437396 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362892 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437392 | - | - |
| Mouse Genome 430 2.0 Array | 1416765_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1416765_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600008973.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008973.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008973.mm.1 | TC1600001133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008975.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008975.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008975.mm.1 | TC1600001133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008979.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008979.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008979.mm.1 | TC1600001133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008978.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008978.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008978.mm.1 | TC1600001133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008982.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008982.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008982.mm.1 | TC1600001133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008974.mm.1 | NM_025571.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008974.mm.1 | NM_025571.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008974.mm.1 | TC1600001133.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416765_PM_s_at | - | - |
| Mouse Expression Set 430 | 1416765_s_at | - | - |
| Murine Genome U74v2 Set | 160258_PM_at | - | - |
| Murine Genome U74v2 Set | 160258_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416765_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416765_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 899999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 580064 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4920452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618461 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243772 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243772 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618461 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001814.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl48807.5.1_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580064 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48807.5.1_32-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001814.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920452 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2976867 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618461 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618461 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2976867 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001814.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl48807.5.1_32-S | - | - |

Pam16
Genomic Map