| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700017249.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017249.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017249.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017253.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017253.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017253.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005253.mm.1 |
NM_001142957.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005253.mm.1 |
NM_001142957.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005253.mm.1 |
TC1700000580.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005252.mm.1 |
NM_001142957.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005252.mm.1 |
NM_001142957.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005252.mm.1 |
TC1700000580.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017247.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017247.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017247.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017251.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017251.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017251.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005248.mm.1 |
NM_001142957.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005248.mm.1 |
NM_001142957.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005248.mm.1 |
TC1700000580.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005254.mm.1 |
NM_001142957.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005254.mm.1 |
NM_001142957.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005254.mm.1 |
TC1700000580.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017248.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017248.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017248.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017250.mm.1 |
NM_029952.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017250.mm.1 |
NM_029952.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017250.mm.1 |
TC1700001864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005255.mm.1 |
NM_001142957.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005255.mm.1 |
NM_001142957.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005255.mm.1 |
TC1700000580.mm.1 |
MTA TC |