| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000011344.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011344.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011344.mm.1 |
TC1000003221.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000024051.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000024051.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000024051.mm.1 |
TC1000003221.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011348.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011348.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011348.mm.1 |
TC1000003221.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011349.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011349.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011349.mm.1 |
TC1000003221.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011347.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011347.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011347.mm.1 |
TC1000003221.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011350.mm.1 |
NM_009040.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011350.mm.1 |
NM_009040.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011350.mm.1 |
TC1000003221.mm.1 |
MTA TC |