Assay Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207301.2 | NP_997184.1 | 5 | 741 | 101 | |
| GenBank mRNA | AK172079.1 | - | 5 | 754 | 101 | |
| BC031769.1 | - | 5 | 741 | 4235618 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wrb
Genomic Map