Assay Details
| Gene Symbol: Acnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145368.2 | NP_663343.2 | 5 | 1417 | 95 | |
| GenBank mRNA | AK050308.1 | - | 5 | 1417 | 95 | |
| AK050357.1 | - | 6 | 1414 | 95 | ||
| BC010829.1 | - | 3 | 1261 | 4220620 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
thiolester hydrolase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
thiolester hydrolase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165091 | NM_145368.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165091 | AK050308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165091 | AK050357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165091 | BC010829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075263 | NM_145368.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075263 | AK050308.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130465 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075263 | NM_145368.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075263 | AK050308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165091 | NM_145368.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165091 | AK050308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165091 | AK050357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165091 | BC010829.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22905080 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05231658 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05231598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130465 | - | - |
| Whole Mouse Genome G4122A | A_51_P213515 | NM_145368.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213515 | AK050308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213515 | AK050357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213515 | BC010829.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127828 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128320 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05231598 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22904975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03266043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04047638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512890 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425150_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512886 | - | - |
| Mouse Genome 430 2.0 Array | 1425150_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425150_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209186_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209186_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400021435.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021435.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021435.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021433.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021433.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021433.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021441.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021441.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021441.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021442.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021442.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021442.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021434.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021434.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021434.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021438.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021438.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021438.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021432.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021432.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021432.mm.1 | NM_145368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021436.mm.1 | TC0400002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021436.mm.1 | NM_145368.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021436.mm.1 | NM_145368.2 | Refseq |
| Mouse Expression Set 430 | 1425150_PM_at | - | - |
| Mouse Expression Set 430 | 1425150_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425150_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425150_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425150_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425150_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 460647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2709810 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2709810 | - | - |
| mouse-6 v1.1 (Illumina) | scl00209186.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00209186.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2709810 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2858121 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2709810 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2858121 | - | - |

Acnat2
Genomic Map