Assay Details
| Gene Symbol: Dusp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023173.2 | NP_075662.2 | 1 | 149 | 134 | |
| GenBank mRNA | AK081862.1 | - | 68 | 134 | ||
| BC099453.1 | - | 1 | 149 | 30938295 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC099453.1, NM_023173.2 transcripts.
Set Membership: |
|

Dusp12
Genomic Map