Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 16 - 17 | 1899 | 77 | |
| GenBank mRNA | AK131964.1 | - | 16 - 17 | 1920 | 77 | |
| BC021755.1 | - | 15 - 16 | 3644 | 77 | ||
| BC052027.1 | - | 16 - 17 | 1926 | 5718637 | 77 | |
| D63784.1 | - | 16 - 17 | 1898 | 77 | ||
| U53208.1 | - | 16 - 17 | 1851 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnajc2
Genomic Map