Assay Details
| Gene Symbol: Sod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011434.1 | NP_035564.1 | 1 - 2 | 190 | 85 | |
| GenBank mRNA | AK020624.1 | - | 1 - 2 | 96 | 85 | |
| AK077284.1 | - | 1 - 2 | 130 | 85 | ||
| BC002066.1 | - | 1 - 2 | 144 | 3496016 | 85 | |
| BC032986.1 | - | 4 - 5 | 85 | |||
| BC048874.1 | - | 1 - 2 | 190 | 5401293 | 85 | |
| BC086886.1 | - | 1 - 2 | 182 | 6760550 | 85 | |
| M35725.1 | - | 1 - 2 | 78 | 85 | ||
| X06683.1 | - | 1 - 2 | 78 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sod1
Genomic Map