Assay Details
| Gene Symbol: BC016548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC016548.1 | - | 1 - 2 | 37 | 59 | |
| BC096650.1 | - | 1 - 2 | 48 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC016548| Gene Symbol: BC016548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC016548.1 | - | 1 - 2 | 37 | 59 | |
| BC096650.1 | - | 1 - 2 | 48 | 59 | ||
Set Membership: |
|