Assay Details
| Gene Symbol: Skint10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177668.2 | NP_808336.1 | 4 - 5 | 636 | 99 | |
| GenBank mRNA | AK037152.1 | - | 4 - 5 | 601 | 99 | |
| EF494905.1 | - | 4 - 5 | 599 | 99 | ||
| EF494906.1 | - | 4 - 5 | 599 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Skint10
Genomic Map