Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112798.2 | NP_001106269.1 | 9 | 17379 | 69 | |
| NM_001286684.1 | NP_001273613.1 | 12 | 14653 | 69 | ||
| NM_011406.3 | NP_035536.2 | 11 | 17415 | 69 | ||
| NR_104580.1 | 9 | 17414 | 69 | |||
| GenBank mRNA | AK046123.1 | - | 1 | 56 | 69 | |
| AK078052.1 | - | 1 | 2446 | 69 | ||
| BC060264.1 | - | 1 | 1396 | 69 | ||
| BC094585.1 | - | 1 | 1630 | 69 | ||
| BC096032.1 | - | 2 | 1244 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc8a1
Genomic Map