Assay Details
| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198028.3 | NP_932145.3 | 1 - 2 | 90 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_198028.3 transcripts.
Set Membership: |
|

Serpinb10
Genomic Map