Assay Details
| Gene Symbol: Dagla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198114.2 | NP_932782.2 | 9 - 10 | 1074 | 122 | |
| GenBank mRNA | AK147384.1 | - | 9 - 10 | 1032 | 122 | |
| AY275376.1 | - | 8 - 9 | 974 | 122 | ||
| BC112414.1 | - | 8 - 9 | 973 | 122 | ||
| BC148308.2 | - | 8 - 9 | 974 | 40061146 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dagla
Genomic Map