Assay Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098222.1 | NP_001091692.1 | 5 | 2669 | 171 | |
| NM_198161.2 | NP_937804.1 | 5 | 2679 | 171 | ||
| GenBank mRNA | AK012577.1 | - | 1 | 1163 | 171 | |
| BC059871.1 | - | 1 | 2034 | 6809364 | 171 | |
| BP760224.1 | - | 1 | 171 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678565 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019407 | AK157023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019407 | BC059871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019407 | NM_198161.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019407 | AK143861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019407 | NM_001098222.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878239 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111838 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019407 | AK157023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019407 | BC059871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019407 | NM_198161.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019407 | AK143861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019407 | NM_001098222.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P323081 | BC059871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P323081 | BP760224.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P323081 | NM_198161.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P323081 | AK012577.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P323081 | NM_001098222.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32678575 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111838 | - | - |
| Whole Mouse Genome G4122A | A_51_P323081 | BC059871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323081 | BP760224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323081 | NM_198161.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323081 | AK012577.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323081 | NM_001098222.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121867 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111838 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085418 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085422 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15085413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678565 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32678566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32678579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15171833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14639694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15073024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14748168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15085423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15085424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14725710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15085422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14748165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32678573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14710009 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601845 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428512_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601844 | - | - |
| Mouse Genome 430 2.0 Array | 1428512_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428512_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70237_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70237_PM_at | - | - |
| Mouse Expression Set 430 | 1428512_at | - | - |
| Mouse Expression Set 430 | 1428512_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428512_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428512_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 549233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1780162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592886 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592886 | - | - |
| mouse-6 v1.1 (Illumina) | scl54620.5_512-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780162 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54620.5_512-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251595 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251595 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54620.5_512-S | - | - |

Bhlhb9
Genomic Map