Assay Details
| Gene Symbol: Ntf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198190.1 | NP_937833.1 | 1 - 2 | 100 | 110 | |
| GenBank mRNA | BC052191.1 | - | 1 - 2 | 100 | 6505258 | 110 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052191.1, NM_198190.1 transcripts.
Set Membership: |
|

Ntf5
Genomic Map