Assay Details
| Gene Symbol: Gtf2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164255.1 | - | 4 - 5 | 769 | 136 | |
| BC048705.1 | - | 3 - 4 | 542 | 6505667 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2a2
Genomic Map