Assay Details
Gene Symbol: Tas2r130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199156.1 | NP_954607.1 | 1 | 845 | 129 | |
GenBank mRNA | BC119329.1 | - | 1 | 927 | 8734078 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
bitter taste receptor activity
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
bitter taste receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156643 | NM_199156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156643 | BC119329.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117894 | NM_199156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117894 | BC119329.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841353 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130867 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168419 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156643 | NM_199156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156643 | BC119329.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117894 | NM_199156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117894 | BC119329.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851540 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851515 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851530 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851539 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851522 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168419 | - | - |
Whole Mouse Genome G4122A | A_52_P259365 | NM_199156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P259365 | BC119329.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168419 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05882589 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05882600 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05674446 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05882597 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05882587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05300939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05054433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05146645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05882585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05835502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05722986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05882601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05674445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05882602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851523 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851528 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10548604 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548603 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548605 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548602 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600025742.mm.1 | TC0600003271.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025742.mm.1 | NM_199156.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025742.mm.1 | NM_199156.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025741.mm.1 | TC0600003271.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025743.mm.1 | TC0600003271.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 379971 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5690053 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005690053 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642480 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2642480 | - | - |
mouse-6 v1.1 (Illumina) | scl0387355.1_236-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5690053 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387355.1_236-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690053 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2642480 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642480 | - | - |
