Assay Details
| Gene Symbol: Tas2r104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207011.1 | NP_996894.1 | 1 | 853 | 74 | |
| GenBank mRNA | BC148229.1 | - | 1 | 857 | 40142081 | 74 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109424 | NM_207011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109424 | BC148229.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150181 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109424 | NM_207011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109424 | BC148229.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851673 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150181 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05689029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05622270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05622277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24851676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24851667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24851669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24851675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24851672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548611 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548610 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025748.mm.1 | TC0600003274.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025748.mm.1 | NM_207011.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025748.mm.1 | NM_207011.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4050575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730744 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730744 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387340.1_274-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387340.1_274-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2730744 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730744 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0387340.1_274-S | - | - |

Tas2r104
Genomic Map