Assay Details
| Gene Symbol: Tas2r113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207018.1 | NP_996901.1 | 1 | 867 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067700 | NM_207018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067700 | NM_207018.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655430 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655450 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655441 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655449 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05655436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05793150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05273496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05610661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05728125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542256 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542255 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012622.mm.1 | TC0600001567.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012622.mm.1 | NM_207018.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012622.mm.1 | NM_207018.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1660040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1660040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722806 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2722806 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387345.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387345.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2722806 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2722806 | - | - |

Tas2r113
Genomic Map