Assay Details
| Gene Symbol: Tas2r121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207024.1 | NP_996907.1 | 1 | 869 | 102 | |
| GenBank mRNA | BC104410.1 | - | 1 | 869 | 40050579 | 102 |
| BC104411.1 | - | 1 | 869 | 40050580 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytokinesis
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
bitter taste receptor activity
detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytokinesis
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
bitter taste receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018452 | BC104410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018452 | BC104411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018452 | NM_207024.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152571 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132469 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018452 | BC104410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018452 | BC104411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018452 | NM_207024.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132469 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05518166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05711454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05502084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05711450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854310 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548664 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548663 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025826.mm.1 | TC0600003285.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025826.mm.1 | NM_207024.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025826.mm.1 | NM_207024.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6220519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607229 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2607229 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387349.1_257-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387349.1_257-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2607229 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2607229 | - | - |

Tas2r121
Genomic Map