Assay Details
| Gene Symbol: Olfr845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207145.2 | NP_997028.1 | 1 | 671 | 98 | |
| GenBank mRNA | BC127982.1 | - | 1 | 672 | 40135240 | 98 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100544 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176179 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339675 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100544 | - | - |
| Whole Mouse Genome G4122A | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510932 | BC127982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P510932 | NM_207145.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176179 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136430 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08119520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07667255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08022121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07667256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08119517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07669612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08119517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26339704 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583395 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583394 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900000975.mm.1 | NM_207145.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000975.mm.1 | NM_207145.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000975.mm.1 | TC0900000148.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700281 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700281 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258249.1_33-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258249.1_33-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2700281 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2700281 | - | - |

Olfr845
Genomic Map