Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700017154.mm.1 |
NM_207245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017154.mm.1 |
NM_207245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017154.mm.1 |
TC1700001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017155.mm.1 |
NM_207245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017155.mm.1 |
NM_207245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017155.mm.1 |
TC1700001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017156.mm.1 |
NM_207245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017156.mm.1 |
NM_207245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017156.mm.1 |
TC1700001855.mm.1 |
MTA TC |