Assay Details
Gene Symbol: Zfp870 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207245.2 | NP_997128.1 | 3 - 4 | 351 | 117 | |
GenBank mRNA | BC066107.1 | - | 3 - 4 | 275 | 30537752 | 117 |
CD350862.1 | - | 3 - 4 | 351 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119058 | NM_207245.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119058 | AK053506.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119058 | BC066107.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495521 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779179 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666666 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133037 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137443 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115965 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119058 | NM_207245.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119058 | AK053506.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119058 | BC066107.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147464 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115965 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03308528 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137443 | - | - |
Whole Mouse Genome G4122A | A_52_P4066 | NM_207245.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P4066 | BC066107.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P232300 | NM_207245.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P232300 | BC066107.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119058 | NM_207245.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119058 | AK053506.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119058 | BC066107.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114908 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101958 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115965 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137443 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13459061 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13459052 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13409930 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13459054 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13459059 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13459058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13130054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13459055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13333707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147448 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13333706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13189902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147472 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13091445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13068939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13276250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13068585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13226455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13276251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147465 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10449939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449938 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455621_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10449935 | - | - |
Mouse Genome 430 2.0 Array | 1455621_at | - | - |
Mouse Genome 430 2.0 Array | 1455621_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240066_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240066_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700017151.mm.1 | NM_207245.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017151.mm.1 | NM_207245.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017151.mm.1 | TC1700001855.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017153.mm.1 | NM_207245.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017153.mm.1 | NM_207245.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017153.mm.1 | TC1700001855.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017152.mm.1 | NM_207245.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017152.mm.1 | NM_207245.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017152.mm.1 | TC1700001855.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455621_PM_at | - | - |
Mouse Expression Set 430 | 1455621_at | - | - |
Murine Genome U74v2 Set | 133802_PM_at | - | - |
Murine Genome U74v2 Set | 133802_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455621_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455621_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 299661 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2650373 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2650373 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002650373 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246614 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246614 | - | - |
mouse-6 v1.1 (Illumina) | scl0240066.1_321-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2650373 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0240066.1_321-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650373 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2903889 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2903889 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0240066.1_321-S | - | - |
