Assay Details
| Gene Symbol: Serpinb1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173051.2 | NP_766639.2 | 4 - 5 | 459 | 147 | |
| GenBank mRNA | BC104332.2 | - | 3 - 4 | 442 | 40045807 | 147 |
More Information
Set Membership: |
|

Serpinb1c
Genomic Map