Assay Details
| Gene Symbol: Serpinb1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173051.2 | NP_766639.2 | 5 - 6 | 596 | 145 | |
| GenBank mRNA | BC104332.2 | - | 4 - 5 | 579 | 40045807 | 145 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859193 | AK032596.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859193 | AK036247.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859193 | NM_029101.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103513 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115340 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174857 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212057 | BC104332.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212057 | NM_173051.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28931929 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07830758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174857 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115340 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07830757 | - | - |
| Whole Mouse Genome G4122A | A_51_P212057 | BC104332.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P212057 | NM_173051.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P493477 | BC104332.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P493477 | NM_173051.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127067 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174857 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115340 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10678658 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10678659 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10791346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10791353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10791339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28931859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10791344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10909193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10553945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11061175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10857724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10791359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28931928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408569 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408565 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010972.mm.1 | NM_173051.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010972.mm.1 | NM_173051.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010972.mm.1 | TC1300001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010973.mm.1 | NM_173051.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010973.mm.1 | NM_173051.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010973.mm.1 | TC1300001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010976.mm.1 | NM_173051.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010976.mm.1 | NM_173051.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010976.mm.1 | TC1300001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010974.mm.1 | NM_173051.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010974.mm.1 | NM_173051.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010974.mm.1 | TC1300001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010975.mm.1 | NM_173051.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010975.mm.1 | NM_173051.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010975.mm.1 | TC1300001764.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2360411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254229 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254229 | - | - |
| mouse-6 v1.1 (Illumina) | scl0380839.1_71-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0380839.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254229 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254229 | - | - |

Serpinb1c
Genomic Map