Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500033040.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033040.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033040.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033036.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033036.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033036.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033034.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033034.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033034.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033035.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033035.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033035.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033042.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033042.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033042.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033037.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033037.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033037.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033041.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033041.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033041.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033039.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033039.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033039.mm.1 |
NM_177380.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033038.mm.1 |
TC0500003599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033038.mm.1 |
NM_177380.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033038.mm.1 |
NM_177380.3 |
Refseq |