Assay Details
| Gene Symbol: Ifna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206871.1 | NP_996754.1 | 1 | 239 | 152 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6329 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16160 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133474 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132609 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115855 | - | - |
| Whole Mouse Genome G4122A | A_51_P192930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23092359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23092387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10505885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514290 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505892 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505888 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505879 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505884 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10514289 | - | - |
| Mouse Genome 430 2.0 Array | 1450593_at | - | - |
| Mouse Genome 430 2.0 Array | 1450593_PM_at | - | - |
| Mouse Expression Set 430 | 1450593_at | - | - |
| Mouse Expression Set 430 | 1422404_x_at | - | - |
| Mouse Expression Set 430 | 1422403_PM_at | - | - |
| Mouse Expression Set 430 | 1450593_PM_at | - | - |
| Mouse Expression Set 430 | 1422403_at | - | - |
| Mouse Expression Set 430 | 1422404_PM_x_at | - | - |
| Murine Genome U74v2 Set | 102148_f_at | - | - |
| Murine Genome U74v2 Set | 102149_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96419_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101788_f_at | - | - |
| Murine Genome U74v2 Set | 102149_f_at | - | - |
| Murine Genome U74v2 Set | 101619_at | - | - |
| Murine Genome U74v2 Set | 102148_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101619_PM_at | - | - |
| Murine Genome U74v2 Set | 101788_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96419_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450593_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450593_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105900593 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103060333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242716 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769622 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257531 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242716 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769622 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230562 | - | - |
| mouse-6 v1.1 (Illumina) | scl0384100.1_5-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_46047422-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840270 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103060333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0384100.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2845312 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242716 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2845312 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769622 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230562 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242716 | - | - |

Ifna6
Genomic Map