Assay Details
Gene Symbol: Ighg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134155.1 | - | 4 - 5 | 1373 | 74 | |
AK156420.1 | - | 5 - 6 | 1065 | 74 | ||
BC010327.1 | - | 6 - 7 | 1149 | 2651776 | 74 | |
BC025447.1 | - | 6 - 7 | 1151 | 5068622 | 74 | |
BC092049.1 | - | 7 - 8 | 1142 | 3661511 | 74 | |
BC092269.1 | - | 6 - 7 | 1131 | 4015795 | 74 | |
BG071858.2 | - | 1 - 2 | 74 | |||
X67210.1 | - | 6 - 7 | 1130 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117903 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066173 | D29669.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066173 | Z37502.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021089 | X70423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021089 | V00798.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021089 | AB097847.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021089 | AJ294738.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021084 | AK135975.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117903 | - | - |
Whole Mouse Genome G4122A | A_51_P117903 | - | - |
Whole Mouse Genome G4122A | A_52_P74106 | BG071858.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P74106 | BC025447.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P74106 | BC092049.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P74106 | BC010327.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P74106 | BC092269.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | X80970.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | X80972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | U88684.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | X80970.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | X80972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P275642 | U88684.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09784637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09873309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28755960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10180176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28755958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10088387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09873322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756083 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10180207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10180203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10076296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28755961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10088396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28756076 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10402874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402942 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402918 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403010 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403055 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402899 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402898 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403075 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402869 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402872 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402873 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424631_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451632_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403009 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10402864 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403073 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403054 | - | - |
Mouse Genome 430 2.0 Array | 1426174_s_at | - | - |
Mouse Genome 430 2.0 Array | 1426174_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1424631_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1424631_a_at | - | - |
Mouse Expression Set 430 | 1424631_PM_a_at | - | - |
Mouse Expression Set 430 | 1426174_PM_s_at | - | - |
Mouse Expression Set 430 | 1451949_at | - | - |
Mouse Expression Set 430 | 1451632_a_at | - | - |
Mouse Expression Set 430 | 1451949_PM_at | - | - |
Mouse Expression Set 430 | 1426174_s_at | - | - |
Mouse Expression Set 430 | 1451632_PM_a_at | - | - |
Mouse Expression Set 430 | 1424631_a_at | - | - |
Murine Genome U74v2 Set | 102824_PM_g_at | - | - |
Murine Genome U74v2 Set | 102153_PM_at | - | - |
Murine Genome U74v2 Set | 98776_at | - | - |
Murine Genome U74v2 Set | 102823_at | - | - |
Murine Genome U74v2 Set | 102844_at | - | - |
Murine Genome U74v2 Set | 102823_PM_at | - | - |
Murine Genome U74v2 Set | 102825_at | - | - |
Murine Genome U74v2 Set | 102159_PM_at | - | - |
Murine Genome U74v2 Set | 102159_at | - | - |
Murine Genome U74v2 Set | 102825_PM_at | - | - |
Murine Genome U74v2 Set | 102153_at | - | - |
Murine Genome U74v2 Set | 102843_PM_s_at | - | - |
Murine Genome U74v2 Set | 102824_g_at | - | - |
Murine Genome U74v2 Set | 102843_s_at | - | - |
Murine Genome U74v2 Set | 99421_PM_at | - | - |
Murine Genome U74v2 Set | 98776_PM_at | - | - |
Murine Genome U74v2 Set | 99421_at | - | - |
Murine Genome U74v2 Set | 102844_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451632_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451949_at | - | - |
Mouse Genome 430A 2.0 Array | 1426174_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1426174_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1424631_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451949_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424631_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451632_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424631_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424631_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249975 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222531 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249975 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222531 | - | - |
