Assay Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291028.1 | NP_001277957.1 | 3 - 4 | 426 | 57 | |
| GenBank mRNA | BC058280.1 | - | 2 - 3 | 350 | 6529102 | 57 |
| DN175861.1 | - | 3 - 4 | 375 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lzts3
Genomic Map