Assay Details
| Gene Symbol: Gm9955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020839.1 | - | 1 | 919 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | formin homology 2 domain containing 3 | Fhod3 |
A930009H06Rik, FHOS2, mKIAA1695 |
Anti-sense | Chr. 18 - 24708456 - 25133507 on NCBI Build GRCm38 | |
| Downstream | tubulin polyglutamylase complex subunit 2 | Tpgs2 |
5730437P09Rik, 5730494M16Rik, AI666318, Pgs2 |
417.9kb | Sense | Chr. 18 - 25127223 - 25169007 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9955
Genomic Map