Assay Details
| Gene Symbol: Mir155hg | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY096003.1 | - | LIT1587 bic |
1 - 2 | 77 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 155 | Mir155 |
Mirn155, mir-155, mmu-mir-155 |
Sense | Chr. 16 - 84714140 - 84714204 on NCBI Build GRCm38 | |
| Downstream | mitochondrial ribosomal protein L39 | Mrpl39 |
C21orf8, MRP-L5, ORF22, Rpml5 |
2.3kb | Anti-sense | Chr. 16 - 84717580 - 84735302 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mir155hg
Genomic Map