Assay Details
| Gene Symbol: Gm10010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039686.1 | - | 1 - 2 | 451 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9330102E08 gene | 9330102E08Rik | -4.0kb | Sense | Chr. 6 - 128169700 - 128183803 on NCBI Build GRCm38 | |
| Downstream | TEA domain family member 4 | Tead4 |
ETFR-2, Etfr2, FR-19, Rtef1, TEAD-4, TEF-3, Tcf13r1, Tef3, Tefr, Tefr1, Tefr1a |
26.5kb | Sense | Chr. 6 - 128227143 - 128300813 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm10010
Genomic Map