Assay Details
| Gene Symbol: 4933406P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016707.1 | - | 1 | 1718 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6413 | Mir6413 |
Gm27432, mmu-mir-6413 |
-12.8kb | Sense | Chr. 10 - 20297581 - 20297688 on NCBI Build GRCm38 |
| Downstream | mitochondrial fission regulator 2 | Mtfr2 |
2610016C23Rik, 4933412C16Rik, Dufd1, Fam54a |
34.7kb | Anti-sense | Chr. 10 - 20346989 - 20361669 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933406P04Rik
Genomic Map