Assay Details
| Gene Symbol: 6530403G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018318.1 | - | 1 | 1236 | 139 | |
| AK045988.1 | - | 1 | 1232 | 138 | ||
| AK084809.1 | - | 1 | 1242 | 139 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycoprotein integral membrane 1 | Ginm1 | AA589616 | -10.4kb | Sense | Chr. 10 - 7767947 - 7780946 on NCBI Build GRCm38 |
| Downstream | peptidylprolyl isomerase (cyclophilin)-like 4 | Ppil4 |
3732410E19Rik, 3830425H19Rik, AI788954, AW146233, PPIase |
1.1kb | Anti-sense | Chr. 10 - 7792891 - 7822563 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6530403G13Rik
Genomic Map