Assay Details
| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081204.1 | NP_001074673.1 | 2 - 3 | 171 | 91 | |
| GenBank mRNA | AB253762.1 | - | 2 - 3 | 136 | 91 | |
| AK089440.1 | - | 2 - 3 | 171 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B3glct
Genomic Map