Assay Details
| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081204.1 | NP_001074673.1 | 3 - 4 | 214 | 104 | |
| GenBank mRNA | AB253762.1 | - | 3 - 4 | 179 | 104 | |
| AK089440.1 | - | 3 - 4 | 214 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B3glct
Genomic Map