Assay Details
| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081204.1 | NP_001074673.1 | 10 - 11 | 897 | 95 | |
| GenBank mRNA | AK089440.1 | - | 10 - 11 | 897 | 95 | |
| AK145082.1 | - | 10 - 11 | 872 | 95 | ||
More Information
Set Membership: |
|

B3glct
Genomic Map