Assay Details
Gene Symbol: Gm13262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK085384.1 | - | 1 - 2 | 148 | 107 | |
AK143820.1 | - | 1 - 2 | 176 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK143820.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128839 | AK085384.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128839 | CX763644.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128839 | AW049595.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128839 | AK030878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128839 | NM_027748.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676080 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938310 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824364 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128839 | AK085384.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128839 | CX763644.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128839 | AW049595.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128839 | AK030878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128839 | NM_027748.3 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04425688 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128839 | AK085384.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128839 | CX763644.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128839 | AW049595.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128839 | AK030878.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128839 | NM_027748.3 | mouse_transcriptome |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01747194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01747195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997650 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01440426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01619214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02218824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02004269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01275522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01600037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20997636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10469141 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446646_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1446646_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1446646_at | - | - |
Mouse Expression Set 430 | 1446646_at | - | - |
Mouse Expression Set 430 | 1446646_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446646_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446646_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 731274 | - | - |
