Assay Details
| Gene Symbol: A130006I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015028.1 | - | 1 | 1051 | 87 | |
| AK079472.1 | - | 1 | 848 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
prenyl (solanesyl) diphosphate synthase, subunit 1 |
Pdss1 |
2610203G20Rik, 2700031G06Rik, TPT, TPT 1, Tprt, mDLP1, mSPS1 |
Anti-sense | Chr. 2 - 22893560 - 22940266 on NCBI Build GRCm38 | |
| Downstream | abl-interactor 1 | Abi1 |
E3B1, NAP1, Ssh3bp1 |
44.5kb | Sense | Chr. 2 - 22939989 - 23040241 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A130006I12Rik
Genomic Map