Assay Details
Gene Symbol: 4933411E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016767.1 | - | 1 - 2 | 279 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2900079G21 gene | 2900079G21Rik |
C130083B14, Gm6179 |
-41.2kb | Anti-sense | Chr. 9 - 112234277 - 112348925 on NCBI Build GRCm38 |
Downstream |
predicted gene, 36367 |
Gm36367 | 826.1kb | Anti-sense | Chr. 9 - 113287712 - 113310985 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305430 | AK016767.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771217 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535695 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375156 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864003 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671212 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423940 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | AB057418.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | AF179302.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | AK048838.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | BC006721.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | BC058093.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568851 | NM_019570.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK002918.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK005034.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK012184.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK029412.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK144721.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK146279.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | AK159723.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | BC028924.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | BC037637.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915948 | NM_025298.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863526 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546933 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281662 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861863 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531391 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773208 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110798 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15818 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930181 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930162 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930238 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930270 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930208 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06930173 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116874 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06930238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07492310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07754038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26835281 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432370_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432370_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432370_at | - | - |
Mouse Expression Set 430 | 1432370_at | - | - |
Mouse Expression Set 430 | 1432370_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432370_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432370_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 482821 | - | - |
