Assay Details
| Gene Symbol: Gm12289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076620.1 | - | 298 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9630013K17 gene | 9630013K17Rik | -44.4kb | Sense | Chr. 11 - 63964535 - 63968209 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2810001G20 gene | 2810001G20Rik | AW045514 | 65.8kb | Sense | Chr. 11 - 64079484 - 64083259 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12289
Genomic Map