Assay Details
| Gene Symbol: Gm9898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087209.1 | - | 1 - 2 | 511 | 99 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104338 | AK087209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104338 | BB534251.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175353 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104338 | AK087209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104338 | BB534251.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175353 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104338 | AK087209.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104338 | BB534251.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00691053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00515577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20120999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00515592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20120996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00045366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00224216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00224217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20120995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00515586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20121022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1459084_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459084_at | - | - |
| Mouse Genome 430 2.0 Array | 1459084_PM_at | - | - |
| Mouse Expression Set 430 | 1459084_at | - | - |
| Mouse Expression Set 430 | 1459084_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459084_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459084_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 699954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104610358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231361 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104610358 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104610358 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231361 | - | - |

Gm9898
Genomic Map