Assay Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199293.1 | NP_001186222.1 | 3 - 4 | 379 | 82 | |
| NM_031386.2 | NP_113563.2 | 3 - 4 | 360 | 82 | ||
| GenBank mRNA | AY193717.1 | - | 3 - 4 | 360 | 82 | |
| AY193718.1 | - | 3 - 4 | 379 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex14
Genomic Map