Assay Details
Gene Symbol: Gm11541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007584.2 | NP_001007585.1 | 1 - 2 | 419 | 95 | |
GenBank mRNA | AK082500.1 | - | 1 - 2 | 420 | 95 | |
BC147833.1 | - | 1 - 2 | 420 | 9087550 | 95 | |
BC147838.1 | - | 1 - 2 | 420 | 9087555 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000003600.rn
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130444 | NM_001007584.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130444 | AK082500.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130444 | BC147833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130444 | BC147838.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139870 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251468 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127806 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697689 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113811 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1774 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169628 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130444 | NM_001007584.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130444 | AK082500.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130444 | BC147833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130444 | BC147838.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034415 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034408 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07448490 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07448499 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169628 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130444 | NM_001007584.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130444 | AK082500.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130444 | BC147833.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130444 | BC147838.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07448490 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169628 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09360017 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09360016 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09360021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09281627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09237314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09281620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09684799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09360019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09684803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09455855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09451334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09362929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09280921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28034419 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390074 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390073 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390072 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100032721.mm.1 | NM_001007584.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032721.mm.1 | NM_001007584.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032721.mm.1 | TC1100003571.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032723.mm.1 | NM_001007584.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032723.mm.1 | NM_001007584.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032723.mm.1 | TC1100003571.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032722.mm.1 | NM_001007584.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032722.mm.1 | NM_001007584.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032722.mm.1 | TC1100003571.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 317152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 100450048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100450048 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231459 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100450048 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100450048 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2883500 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883500 | - | - |
