Assay Details
| Gene Symbol: 2310034O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040679.1 | 3 - 4 | 506 | 72 | ||
| GenBank mRNA | AK009622.1 | - | 3 - 4 | 506 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 150C | Tmem150c | -51.0kb | Anti-sense | Chr. 5 - 100077872 - 100165604 on NCBI Build GRCm38 | |
| Downstream | Sec31 homolog A (S. cerevisiae) | Sec31a |
1810024J13Rik, ABP125, ABP130, AU042160, HSPC275, Sec31l1 |
143.6kb | Anti-sense | Chr. 5 - 100361649 - 100416234 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2310034O05Rik
Genomic Map