Assay Details
| Gene Symbol: Rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005525.2 | NP_001005525.1 | 5 - 6 | 715 | 92 | |
| GenBank mRNA | AK005213.1 | - | 3 - 4 | 446 | 92 | |
| AK012697.1 | - | 6 - 7 | 782 | 92 | ||
| AK054360.1 | - | 2 - 3 | 1285 | 92 | ||
| AK084745.1 | - | 6 - 7 | 762 | 92 | ||
| AK166941.1 | - | 8 - 9 | 953 | 92 | ||
| BC008229.1 | - | 5 - 6 | 673 | 92 | ||
| BC132550.1 | - | 3 - 4 | 499 | 40130827 | 92 | |
| BC132552.1 | - | 3 - 4 | 499 | 40130829 | 92 | |
| BC145430.1 | - | 3 - 4 | 498 | 9053988 | 92 | |
| BC145431.1 | - | 3 - 4 | 499 | 92 | ||
| BC145432.1 | - | 3 - 4 | 499 | 92 | ||
| DT902145.1 | - | 1 - 2 | 178 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rsrc2
Genomic Map