Assay Details
| Gene Symbol: Rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005525.2 | NP_001005525.1 | 8 - 9 | 1162 | 132 | |
| GenBank mRNA | AA168900.1 | - | 2 - 3 | 378 | 132 | |
| AK005213.1 | - | 6 - 7 | 897 | 132 | ||
| BC132550.1 | - | 6 - 7 | 946 | 40130827 | 132 | |
| BC132552.1 | - | 6 - 7 | 946 | 40130829 | 132 | |
| BC145430.1 | - | 6 - 7 | 942 | 9053988 | 132 | |
| BC145431.1 | - | 7 - 8 | 1021 | 132 | ||
| BC145432.1 | - | 6 - 7 | 941 | 132 | ||
| CN525339.1 | - | 2 - 3 | 295 | 132 | ||
| DT902145.1 | - | 4 - 5 | 625 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rsrc2
Genomic Map