Assay Details
| Gene Symbol: A930006L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044309.1 | - | 1 - 2 | 1060 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 38 | Zbtb38 |
A930014K01Rik, CIBZ |
Anti-sense | Chr. 9 - 96685393 - 96731675 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA E030011O05 gene | E030011O05Rik | 20.1kb | Anti-sense | Chr. 9 - 96731575 - 96752831 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

A930006L05Rik
Genomic Map