Assay Details
Gene Symbol: 5430431A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017368.1 | - | 1 - 2 | 479 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Josephin domain containing 2 | Josd2 | 1110007C05Rik | -12.9kb | Sense | Chr. 7 - 44467980 - 44471658 on NCBI Build GRCm38 |
Downstream | ER membrane protein complex subunit 10 | Emc10 |
2310044H10Rik, 5430410O10Rik, Inm02, Mirta22 |
4.5kb | Anti-sense | Chr. 7 - 44489938 - 44496524 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
