Assay Details
| Gene Symbol: 4833419F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040328.1 | 3 - 4 | 549 | 64 | ||
| GenBank mRNA | AK014726.1 | - | 3 - 4 | 551 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase 8 | Map3k8 |
Cot, Cot/Tpl2, Est, Estf, Tpl-2, Tpl2, c-COT |
-14.4kb | Anti-sense | Chr. 18 - 4331325 - 4353481 on NCBI Build GRCm38 |
| Downstream | mitochondrial poly(A) polymerase | Mtpap |
0610027A18Rik, AW551379, Papd1 |
6.6kb | Sense | Chr. 18 - 4375592 - 4398800 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4833419F23Rik
Genomic Map